Läs mer
3,5-Dimethoxyphenylacetonitrile, 98+%
Produktkod. 11397766
Change view
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11397766 5 g 5g
11387766 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 11397766 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer L11443.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
CAS: 13388-75-5 | C10H11NO2 | 177.20 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13388-75-5
Molekylformel C10H11NO2
Molekylvikt (g/mol) 177.20
MDL-nummer MFCD00016395
InChI-nyckel UUNRWZQWCNTSCV-UHFFFAOYSA-N
Synonym 3,5-dimethoxyphenylacetonitrile, 2-3,5-dimethoxyphenyl acetonitrile, 3,5-dimethoxyphenyl acetonitrile, 3,5-dimethoxy-benzeneacetonitrile, benzeneacetonitrile, 3,5-dimethoxy, 2-3,5-dimethoxyphenyl ethanenitrile, pubchem16207, 3,5-dimethoxybenzylcyanide, acmc-20a41u
PubChem CID 139445
IUPAC-namn 2-(3,5-dimethoxyphenyl)acetonitrile
LEDER COC1=CC(OC)=CC(CC#N)=C1
Specifikationer
Kemiskt namn eller material 3,5-Dimethoxyphenylacetonitrile
Smältpunkt 53°C to 56°C
Odör Characteristic
Kvantitet 5 g
Beilstein 2096481
Formel vikt 177.2
Procent renhet ≥98%