missing translation for 'onlineSavingsMsg'
Learn More

3,5,-dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1h-pyrazole, 97%, Thermo Scientific™

Catalog Number p-3586707
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
937796-06-0
C17H23BN2O2
298.19
MFCD09879979
SERWCEBXGSSDRB-UHFFFAOYSA-N
3,5-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole, 4-3,5-dimethylpyrazol-1-yl phenylboronic acid, pinacol ester, 3,5-dimethyl-1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 3,5,-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole, 3,5-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 4-3,5-dimethyl-1h-pyrazol-1-yl benzeneboronic acid,pinacol ester, 3,5-dimethyl-1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole
24229763
3,5-dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrazole
CC1=NN(C(C)=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
937796-06-0
C17H23BN2O2
298.19
MFCD09879979
SERWCEBXGSSDRB-UHFFFAOYSA-N
3,5-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole, 4-3,5-dimethylpyrazol-1-yl phenylboronic acid, pinacol ester, 3,5-dimethyl-1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 3,5,-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole, 3,5-dimethyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 4-3,5-dimethyl-1h-pyrazol-1-yl benzeneboronic acid,pinacol ester, 3,5-dimethyl-1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole
24229763
3,5-dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrazole
CC1=NN(C(C)=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 298.19
Percent Purity 97%
Chemical Name or Material 3,5,-Dimethyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'