Läs mer
3,5-Dimethyl-2-vinylpyrazine, TRC
Produktkod. 30135463
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135463 100 mg
30133255 50 mg
30009234 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135463 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD492896100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 157615-33-3
Molekylformel C8H10N2
Molekylvikt (g/mol) 134.18
Synonym 2-Ethenyl-3,5-dimethylpyrazine
LEDER C=CC1=NC=C(C)N=C1C
Specifikationer
Kemiskt namn eller material 3,5-Dimethyl-2-vinylpyrazine