missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081287
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
499-06-9
C9H10O2
150.177
MFCD00002525
UMVOQQDNEYOJOK-UHFFFAOYSA-N
mesitylenic acid, benzoic acid, 3,5-dimethyl, unii-ed8av34n0y, 3,5-dimethyl benzoic acid, 3,5-dimethyl-benzoic acid, ed8av34n0y, 3,5-dimethylbenzoicacid, pubchem15440, 3,5-dimethylbezoic acid, 3,5 dimethylbenzoic acid
10356
CHEBI:64821
3,5-dimethylbenzoic acid
CC1=CC(=CC(=C1)C(=O)O)C
This item is not returnable. View return policy
499-06-9
C9H10O2
150.177
MFCD00002525
UMVOQQDNEYOJOK-UHFFFAOYSA-N
mesitylenic acid, benzoic acid, 3,5-dimethyl, unii-ed8av34n0y, 3,5-dimethyl benzoic acid, 3,5-dimethyl-benzoic acid, ed8av34n0y, 3,5-dimethylbenzoicacid, pubchem15440, 3,5-dimethylbezoic acid, 3,5 dimethylbenzoic acid
10356
CHEBI:64821
3,5-dimethylbenzoic acid
CC1=CC(=CC(=C1)C(=O)O)C

3,5-Dimethylbenzoic acid is suitable reagent used as carbon and energy supplement in the culture medium of Pseudomonas sp

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylbenzoic acid is suitable reagent used as carbon and energy supplement in the culture medium of Pseudomonas sp

Solubility
Soluble in methanol. (1 g/10 mL). Slightly soluble in water.

Notes
Store away from oxidizing agents. Use only in a chemical fume hood. Incompatible with strong acids and bases.
TRUSTED_SUSTAINABILITY
Melting Point 168°C to 172°C
Quantity 10 g
Odor Odorless
Beilstein 1072182
Solubility Information Soluble in methanol. (1g/10 mL). Slightly soluble in water.
Formula Weight 150.18
Percent Purity ≥98%
Chemical Name or Material 3,5-Dimethylbenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'