missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenyl isothiocyanate, 97%, Maybridge™

512.13 SEK

Chemical Identifiers

CAS 40046-30-8
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.238
MDL Number MFCD00041082
InChI Key DSMXCADWIFIJEX-UHFFFAOYSA-N
Synonym 3,5-dimethylphenyl isothiocyanate, 3,5-dimethylphenylisothiocyanate, 3,5-dimethylbenzenisothiocyanate, acmc-20anca, 3,5-xylyl isothiocyanate, 1-isothiocyanato-3,5-dimethyl-benzene, #, isothiocyanic acid 3,5-dimethylphenyl ester
PubChem CID 142406
IUPAC Name 1-isothiocyanato-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)N=C=S)C
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10019511
SDS View Documents
Maybridge
TL00501EA
10g
512.13 SEK
10g
View alternative products
Save Up to 
»
Estimated Shipment
16-03-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

40046-30-8
163.238
DSMXCADWIFIJEX-UHFFFAOYSA-N
142406
CC1=CC(=CC(=C1)N=C=S)C
C9H9NS
MFCD00041082
3,5-dimethylphenyl isothiocyanate, 3,5-dimethylphenylisothiocyanate, 3,5-dimethylbenzenisothiocyanate, acmc-20anca, 3,5-xylyl isothiocyanate, 1-isothiocyanato-3,5-dimethyl-benzene, #, isothiocyanic acid 3,5-dimethylphenyl ester
1-isothiocyanato-3,5-dimethylbenzene

Specifications

3, 5-Dimethylphenyl isothiocyanate
40046-30-8
97%
(CH3)2C6H3NCS
3,5-dimethylphenyl isothiocyanate, 3,5-dimethylphenylisothiocyanate, 3,5-dimethylbenzenisothiocyanate, acmc-20anca, 3,5-xylyl isothiocyanate, 1-isothiocyanato-3,5-dimethyl-benzene, #, isothiocyanic acid 3,5-dimethylphenyl ester
CC1=CC(=CC(=C1)N=C=S)C
163.238
163.24
10g
97.0
C9H9NS
MFCD00041082
DSMXCADWIFIJEX-UHFFFAOYSA-N
1-isothiocyanato-3,5-dimethylbenzene
142406
97%
SDS