missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrobenzoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog Number p-7023311
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
99-33-2
C7H3ClN2O5
230.56
MFCD00007248
NNOHXABAQAGKRZ-UHFFFAOYSA-N
benzoyl chloride, 3,5-dinitro, 3,5-dinitrobenzoic acid chloride, dnbc, 3,5-dinitrobenzoylchloride, unii-5jfa2dvm4d, ccris 3137, 5jfa2dvm4d, 3,5-dinitrobenzoyl, acmc-209scd, dnbc;
7432
3,5-dinitrobenzoyl chloride
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)Cl
This item is not returnable. View return policy
99-33-2
C7H3ClN2O5
230.56
MFCD00007248
NNOHXABAQAGKRZ-UHFFFAOYSA-N
benzoyl chloride, 3,5-dinitro, 3,5-dinitrobenzoic acid chloride, dnbc, 3,5-dinitrobenzoylchloride, unii-5jfa2dvm4d, ccris 3137, 5jfa2dvm4d, 3,5-dinitrobenzoyl, acmc-209scd, dnbc;
7432
3,5-dinitrobenzoyl chloride
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)Cl

Used in photography. This aromatic compound is used by chemists to identify alcohol components in esters and in the fluorometric analysis of creatinine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in photography. This aromatic compound is used by chemists to identify alcohol components in esters and in the fluorometric analysis of creatinine.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, water.
TRUSTED_SUSTAINABILITY
Melting Point 67°C to 70°C
Boiling Point 196°C (12 mmHg)
Quantity 25 g
UN Number UN3261
Beilstein 990249
Sensitivity Moisture sensitive
Merck Index 14,3277
Solubility Information Slightly soluble in water.
Formula Weight 230.57
Percent Purity ≥98%
Chemical Name or Material 3,5-Dinitrobenzoyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,5-Dinitrobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : 3,5-dinitro-benzoylchloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'