Läs mer
3,5-Dinitrosalicylhydrazide-15N2, TRC
Produktkod. 17941365
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17941365 10 mg
17912235 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17941365 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD48031710MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1346598-09-1
Molekylformel C7H6N215N2O6
Molekylvikt (g/mol) 244.13
Synonym 3,5-Dinitrosalicylic acid 15N-hydrazide, 3,5-Dinitrosalicylhydrazide-15N2, Nifursol-desfurfuryliden-15N2, 2-Hydroxy-3,5-dinitrobenzoic acid hydrazide-15N2, 2-Hydroxy-3,5-dinitrobenzohydrazide-15N2, Nifursol-desfurfuryliden 15N2 (hydrazide 15N2)
IUPAC-namn 2-hydroxy-3,5-dinitro(^{15}N)benzohydrazide
LEDER [15NH2][15NH]C(=O)c1cc(cc(c1O)[N+](=O)[O-])[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 3,5-Dinitrosalicylhydrazide-15N2
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C7H6N4O6/c8-9-7(13)4-1-3(10(14)15)2-5(6(4)12)11(16)17/h1-2,12H,8H2,(H,9,13)/i8+1,9+1
Formel vikt 244.0228
Procent renhet >95%