Läs mer
3,5-Dinitrosalicylic Acid Hydrazide, Mikromol™
Produktkod. 17962896
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17962896 100 mg
17961606 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17962896 Leverantör Mikromol Leverantörsnummer MM0444.03
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3,5-Dinitrosalicylic Acid Hydrazide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,5-Dinitrosalicylic Acid Hydrazide
CAS 955-07-7
Analyt- eller komponentnamn 3,5-Dinitrosalicylic Acid Hydrazide
Frakt skick Room Temperature
Molekylformel C7 H6 N4 O6
Certifikat och efterlevnad ISO 17025
Synonym 3,5-Dinitrosalicylic Acid Hydrazide, 3,5-Dinitrosalicylhydrazide, Nifursol-desfurfuryliden
InChI formel InChI=1S/C7H6N4O6/c8-9-7(13)4-1-3(10(14)15)2-5(6(4)12)11(16)17/h1-2,12H,8H2,(H,9,13)
LEDER NNC(=O)c1cc(cc(c1O)[N+](=O)[O-])[N+](=O)[O-]
IUPAC-namn 2-hydroxy-3,5-dinitrobenzohydrazide
Molekylvikt (g/mol) 242.15
Kvantitet 100 mg
Formel vikt 242.0287 g/mol