Läs mer
3-(5-Nitrofurfurylideneamino)hydantoic Acid, TRC
Produktkod. 17982235
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17982235 1 g
17973295 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17982235 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN4938851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 63981-22-6
Molekylformel C8 H8 N4 O6
Molekylvikt (g/mol) 256.17
Synonym Acetic acid, 2-[1-(aminocarbonyl)-2-[(5-nitro-2-furanyl)methylene]hydrazinyl]-, Acetic acid, [(aminocarbonyl)[(5-nitro-2-furanyl)methylene]hydrazino]- (9CI), Hydantoic acid, 3-(5-nitrofurfurylideneamino)- (6CI), Nitrofurantoin Related Compound A (USP)
IUPAC-namn 2-[carbamoyl-[(5-nitrofuran-2-yl)methylideneamino]amino]acetic acid
LEDER NC(=O)N(CC(=O)O)N=Cc1oc(cc1)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material N-(Aminocarbonyl)-N-[([5-nitro-2-furanyl]-methylene)-amino]-glycine
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C8H8N4O6/c9-8(15)11(4-7(13)14)10-3-5-1-2-6(18-5)12(16)17/h1-3H,4H2,(H2,9,15)(H,13,14)
Formel vikt 256.0444
Procent renhet >95%