Läs mer
3-[(5S)-(4-Fluorophenyl)-5-hydroxypentanoyl]-(4S)-phenyl-1,3-oxazolidin-2-one, TRC
Produktkod. 30405058
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30405058 500 mg
30403240 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30405058 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF595465500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 189028-95-3
Molekylformel C20H20FNO4
Molekylvikt (g/mol) 357.38
Synonym (4S)-4-Phenyl-3-[(5S)-5-(4-fluorophenyl)-5-hydroxypentanoyl]-1,3-oxazolidin-2-one, [S-(R*,R*)]-3-[5-(4-Fluorophenyl)-5-hydroxy-1-oxopentyl]-4-phenyl-2-oxazolidinone, (S)-3-((S)-5-(4-Fluorophenyl)-5-hydroxypentanoyl)-4-phenyloxazolidin-2-one
LEDER O=C(N1[C@@H](C2=CC=CC=C2)COC1=O)CCC[C@@H](C3=CC=C(F)C=C3)O
Specifikationer
Kemiskt namn eller material 3-[(5S)-(4-Fluorophenyl)-5-hydroxypentanoyl]-(4S)-phenyl-1,3-oxazolidin-2-one
Kvantitet 500 mg
InChI formel InChI=1S/C20H20FNO4/c21-16-11-9-15(10-12-16)18(23)7-4-8-19(24)22-17(13-26-20(22)25)14-5-2-1-3-6-14/h1-3,5-6,9-12,17-18,23H,4,7-8,13H2/t17-,18+/m1/s1
Formel vikt 357.1376