Läs mer
3,6-Cyclopropylene-Delta4-tetrahydrophthalic Anhydride, TRC
Produktkod. 30363833
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363833 100 mg
30362162 50 mg
30358182 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30363833 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC988988100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 944-41-2
Molekylformel C11H10O3
Molekylvikt (g/mol) 190.2
Synonym rel-(3aR,4R,4aR,5aS,6S,6aS)-3a,4,4a,5,5a,6-Hexahydro-4,6-etheno-1H-cycloprop[f]isobenzofuran-1,3(3aH)-dione, Tricyclo[3.2.2.02,4]non-8-ene-6,7-dicarboxylic Anhydride Stereoisomer, (3aalpha,4beta,4aalpha,5aalpha,6beta,6aalpha)-4,4a,5,5a,6,6a-Hexahydro-4,6-etheno-1H-cycloprop[f]isobenzofuran-1,3(3aH)-dione
IUPAC-namn (1S,2S,6R,7R,8R,10S)-4-oxatetracyclo[5.3.2.02,6.08,10]dodec-11-ene-3,5-dione
LEDER O=C1OC(=O)[C@H]2[C@H]3C=C[C@H]([C@@H]4C[C@H]34)[C@@H]12
Specifikationer
Kemiskt namn eller material 3,6-Cyclopropylene-delta4-tetrahydrophthalic Anhydride
Kvantitet 100 mg
InChI formel InChI=1S/C11H10O3/c12-10-8-4-1-2-5(7-3-6(4)7)9(8)11(13)14-10/h1-2,4-9H,3H2/t4-,5+,6+,7-,8-,9+
Formel vikt 190.063