Läs mer
3,6-Dimethyl-3-heptanol, TRC
Produktkod. 30375233
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30375233 100 mg
30376654 250 mg
30376551 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30375233 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD473110100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1573-28-0
Molekylformel C9 H20 O
Molekylvikt (g/mol) 144.25
Synonym 3,6-Dimethylheptan-3-ol
IUPAC-namn 3,6-dimethylheptan-3-ol
LEDER CCC(C)(O)CCC(C)C
Specifikationer
Kemiskt namn eller material 3,6-Dimethyl-3-heptanol
Kvantitet 100 mg
InChI formel InChI=1S/C9H20O/c1-5-9(4,10)7-6-8(2)3/h8,10H,5-7H2,1-4H3
Formel vikt 144.1514