Läs mer
3,6-Dioxaoctane-1,8-diyl Bismethanethiosulfonate, TRC
Produktkod. 30379065
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30379065 5 mg
30375208 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30379065 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4861505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 212262-04-9
Molekylformel C8 H18 O6 S4
Molekylvikt (g/mol) 338.48
IUPAC-namn 1,2-bis(2-methylsulfonylsulfanylethoxy)ethane
LEDER CS(=O)(=O)SCCOCCOCCSS(=O)(=O)C
Specifikationer
Kemiskt namn eller material 3,6-Dioxaoctane-1,8-diyl Bismethanethiosulfonate
Kvantitet 5 mg
InChI formel InChI=1S/C8H18O6S4/c1-17(9,10)15-7-5-13-3-4-14-6-8-16-18(2,11)12/h3-8H2,1-2H3
Formel vikt 337.9986