Läs mer
3,6-Methylene-2,5-piperazinedione, TRC
Produktkod. 30473874
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30473874 50 mg
30473757 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30473874 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM30424050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15996-22-2
Molekylformel C6H6N2O2
Molekylvikt (g/mol) 138.12
Synonym 3,6-Bis(methylene)-2,5-piperazinedione, 2,5-Dimethylene-3,6-diketopiperazine, 3,6-Dimethylene-2,5-piperazinedione, NSC 278175
LEDER C=C1NC(=O)C(=C)NC1=O
Specifikationer
Kemiskt namn eller material 3,6-Methylene-2,5-piperazinedione
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C6H6N2O2/c1-3-5(9)8-4(2)6(10)7-3/h1-2H2,(H,7,10)(H,8,9)
Procent renhet >95%