missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,7-Dihydro-4H-imidazo[4,5-d]-1,2,3-triazin-4-one monohydrat (2-azahypoxantin monohydrat), Mikromol™
Description
3,7-Dihydro-4H-imidazo[4,5-d]-1,2,3-triazin-4-one monohydrat (2-azahypoxantin monohydrat), Mikromol
Specifications
Specifications
| Kemiskt namn eller material | 2-Azahypoxanthine Monohydrate |
| CAS | 3-3-7151 |
| Analyt- eller komponentnamn | 2-Azahypoxanthine Monohydrate |
| Frakt skick | Rumstemperatur |
| Molekylformel | C4 H3 N5 O . H2 O |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Dacarbazine Imp. A (EP), Temozolomide Imp. E (EP), Dacarbazine USP Related Compound B, Dacarbazine USP RC B, 3,7-Dihydro-4H-imidazo[4,5-d]-1,2,3-triazin-4-one Monohydrate, 2-Azahypoxanthine Monohydrate, Dacarbazine Related Compound B as Monohydrate, Dacarbazine Impurity A as Monohydrate, Temozolomide Impurity E as Monohydrate |
| InChI formel | InChI=1S/C4H3N5O.H2O/c10-4-2-3(6-1-5-2)7-9-8-4;/h1H,(H2,5,6,7,8,10);1H2 |
| Lösningstyp | Snyggt |
| LEDER | O.O=C1NN=Nc2[nH]cnc12 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?