missing translation for 'onlineSavingsMsg'
Läs mer

3-acetamidoethyl-3-hydroxy-5-methoxyindolin-2-one, TRC

Produktkod. 30337814
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30337814 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337814 Supplier Toronto Research Chemicals Supplier No. TRCA840300250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 3-acetamidoethyl-3-hydroxy-5-methoxyindolin-2-one
CAS 515128-09-3
Molekylformel C13H16N2O4
Kvantitet 250 mg
Synonym N-(2-(3-hydroxy-5-methoxy-2-oxoindolin-3-yl)ethyl)acetamide
InChI formel InChI=1S/C13H16N2O4/c1-8(16)14-6-5-13(18)10-7-9(19-2)3-4-11(10)15-12(13)17/h3-4,7,18H,5-6H2,1-2H3,(H,14,16)(H,15,17)
LEDER O=C(NCCC1(O)C(=O)NC2=CC=C(OC)C=C21)C
IUPAC-namn N-[2-(3-hydroxy-5-methoxy-2-oxo-1H-indol-3-yl)ethyl]acetamide
Molekylvikt (g/mol) 264.28
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.