Läs mer
3-Acetyl Rocuronium Bromide, TRC
Produktkod. 30133746
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133746 500 mg
30010342 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133746 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA187730500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 122483-73-2
Molekylformel C34 H55 N2 O5 . Br
Molekylvikt (g/mol) 651.71
Synonym 1-[3α,17β-Diacetoxy-2β-(morpholin-4-yl)-5α-androstan-16β-yl]-1-(prop-2-enyl)pyrrolidinium Bromide, Pyrrolidinium, 1-[(2β,3α,5α,16β,17β)-3,17-bis(acetyloxy)-2-(4-morpholinyl)androstan-16-yl]-1-(2-propen-1-yl)-, bromide (1:1), Pyrrolidinium, 1-[(2β,3α,5α,16β,17β)-3,17-bis(acetyloxy)-2-(4-morpholinyl)androstan-16-yl]-1-(2-propenyl)-, bromide (9CI)
IUPAC-namn [(2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-17-acetyloxy-10,13-dimethyl-2-morpholin-4-yl-16-(1-prop-2-enylpyrrolidin-1-ium-1-yl)-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate;bromide
LEDER [Br-].CC(=O)O[C@H]1C[C@@H]2CC[C@@H]3[C@H](CC[C@@]4(C)[C@H]3C[C@@H]([C@@H]4OC(=O)C)[N+]5(CC=C)CCCC5)[C@@]2(C)C[C@@H]1N6CCOCC6
Specifikationer
Kemiskt namn eller material 3-Acetyl Rocuronium Bromide
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C34H55N2O5.BrH/c1-6-15-36(16-7-8-17-36)30-21-28-26-10-9-25-20-31(40-23(2)37)29(35-13-18-39-19-14-35)22-34(25,5)27(26)11-12-33(28,4)32(30)41-24(3)38;/h6,25-32H,1,7-22H2,2-5H3;1H/q+1;/p-1/t25-,26+,27-,28-,29-,30-,31-,32-,33-,34-;/m0./s1
Formel vikt 650.3294
Procent renhet >95%