Läs mer
3-(Acetyloxy)-5-iodophenol-2’,3’,4’-tri-O-acetyl-Beta-D-glucuronide Methyl Ester, TRC
Produktkod. 30311342
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30311342 50 mg
1 options
Produktkod. 30311342 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA16600050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-(Acetyloxy)-5-iodophenol-2',3',4'-tri-O-acetyl-beta-D-glucuronide Methyl Ester
CAS 490028-21-2
Molekylformel C21 H23 I O12
Kvantitet 50 mg
Synonym 3-(Acetyloxy)-5-iodophenyl-beta-D-glucopyranosiduronic Acid Methyl Ester Triacetate
InChI formel InChI=1S/C21H23IO12/c1-9(23)29-14-6-13(22)7-15(8-14)33-21-19(32-12(4)26)17(31-11(3)25)16(30-10(2)24)18(34-21)20(27)28-5/h6-8,16-19,21H,1-5H3/t16-,17-,18-,19+,21+/m0/s1
LEDER COC(=O)[C@H]1O[C@@H](Oc2cc(I)cc(OC(=O)C)c2)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC-namn methyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(3-acetyloxy-5-iodophenoxy)oxane-2-carboxylate
Molekylvikt (g/mol) 594.305
Formel vikt 594.023