missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1,2,4-triazole, 96.1%, MP Biomedicals™

Catalog Number p-4403815
Click to view available options
Quantity:
10 g
25 g
100 g
This item is not returnable. View return policy
61-82-5
C2H4N4
84.08
MFCD00005230,MFCD00053362
KLSJWNVTNUYHDU-UHFFFAOYSA-N
3-amino-1,2,4-triazole, amitrole, aminotriazole, 4h-1,2,4-triazol-3-amine, 1h-1,2,4-triazol-3-amine, amitrol, amerol, amizol, emisol, vorox
1639
CHEBI:40036
1H-1,2,4-triazol-5-amine
NC1=NC=NN1
This item is not returnable. View return policy
61-82-5
C2H4N4
84.08
MFCD00005230,MFCD00053362
KLSJWNVTNUYHDU-UHFFFAOYSA-N
3-amino-1,2,4-triazole, amitrole, aminotriazole, 4h-1,2,4-triazol-3-amine, 1h-1,2,4-triazol-3-amine, amitrol, amerol, amizol, emisol, vorox
1639
CHEBI:40036
1H-1,2,4-triazol-5-amine
NC1=NC=NN1
Color White
Density ≥0.6 to 0.85g/cm3 at 20°C
Assay Percent Range 0.961
Quantity 10 g
Formula Weight 84.08
Physical Form Powder
Chemical Name or Material 3-Amino-1, 2, 4-triazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'