Läs mer
3-Amino-1-methyl-5H-pyrido[4,3-b]indole-13C2,15N Acetate, TRC
Produktkod. 30310195
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30310195 10 mg
1 options
Produktkod. 30310195 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA61800210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Amino-1-methyl-5H-pyrido[4,3-b]indole-13C2,15N Acetate
CAS 1216852-93-5
Molekylformel C1213C2H15N215NO2
Kvantitet 10 mg
Synonym 1-Methyl-5H-pyrido[4,3-b]indol-3-amine-13C2,15N Acetate, TRP-P-2-13C2,15N Acetate
InChI formel InChI=1S/C12H11N3.C2H4O2/c1-7-12-8-4-2-3-5-9(8)15-10(12)6-11(13)14-7;1-2(3)4/h2-6,15H,1H3,(H2,13,14);1H3,(H,3,4)/i6+1,11+1,13+1;
LEDER CC1=N[13C]([15NH2])=[13CH]C2=C1C3=C(N2)C=CC=C3.CC(O)=O
IUPAC-namn acetic acid;1-methyl-5H-(5,6-13C2)pyridino[4,3-b]indol-3-(15N)amine
Molekylvikt (g/mol) 260.27