missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-chloro-4-methylpyridine, 97%, Thermo Scientific™

Catalog Number p-7028659
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
133627-45-9
C6H7ClN2
142.59
MFCD00673152
UOBCYTOUXLAABU-UHFFFAOYSA-N
3-amino-2-chloro-4-methylpyridine, 2-chloro-3-amino-4-methylpyridine, 3-amino-2-chloro-4-picoline, 2-chloro-3-amino-4-methyl pyridine, 2-chloro-4-methyl-pyridin-3-ylamine, 2-chloro-3-amino-4-picoline, 2-chloro-4-methyl-3-pyridinamine, 3-pyridinamine, 2-chloro-4-methyl, 2-chloro-4-methyl-3-pyridylamine, 3-amino-2-chloro-4-methyl pyridine
2756387
2-chloro-4-methylpyridin-3-amine
CC1=CC=NC(Cl)=C1N
This item is not returnable. View return policy
133627-45-9
C6H7ClN2
142.59
MFCD00673152
UOBCYTOUXLAABU-UHFFFAOYSA-N
3-amino-2-chloro-4-methylpyridine, 2-chloro-3-amino-4-methylpyridine, 3-amino-2-chloro-4-picoline, 2-chloro-3-amino-4-methyl pyridine, 2-chloro-4-methyl-pyridin-3-ylamine, 2-chloro-3-amino-4-picoline, 2-chloro-4-methyl-3-pyridinamine, 3-pyridinamine, 2-chloro-4-methyl, 2-chloro-4-methyl-3-pyridylamine, 3-amino-2-chloro-4-methyl pyridine
2756387
2-chloro-4-methylpyridin-3-amine
CC1=CC=NC(Cl)=C1N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 64°C to 68°C
Quantity 1 g
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 3-Amino-2-chloro-4-methylpyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'