Läs mer
3'-Amino-2'-hydroxyacetophenone Hydrochloride, TRC
Produktkod. 30325091
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30325091 25 g
1 options
Produktkod. 30325091 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA61118025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3'-Amino-2'-hydroxyacetophenone Hydrochloride
CAS 90005-55-3
Molekylformel C8H10ClNO2
Kvantitet 25 g
Synonym 2-Acetyl-6-aminophenol Hydrochloride, 1-(3-Amino-2-hydroxyphenyl)ethanone Hydrochloride, 3'-Amino-2'-hydroxy-acetophenone Hydrochloride, 1-(3-Amino-2-hydroxyphenyl)ethanone Hydrochloride
InChI formel InChI=1S/C8H9NO2.ClH/c1-5(10)6-3-2-4-7(9)8(6)11;/h2-4,11H,9H2,1H3;1H
LEDER Cl.CC(=O)c1cccc(N)c1O
IUPAC-namn 1-(3-amino-2-hydroxyphenyl)ethanone;hydrochloride
Molekylvikt (g/mol) 187.62
Formel vikt 187.04