Läs mer
3-Amino-4-(2-chlorophenyl)-6-nitro-2(1H)-quinolinone (Clonazepam Impurity), TRC
Produktkod. 17994515
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17994515 10 mg
17993925 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17994515 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA60359010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55198-89-5
Molekylformel C15 H10 Cl N3 O3
Molekylvikt (g/mol) 315.71
Synonym 3-Amino-4-(2-chlorophenyl)-6-nitroquinolin-2(1H)-one, Clonazepam Imp. B (EP)
IUPAC-namn 3-amino-4-(2-chlorophenyl)-6-nitro-1H-quinolin-2-one
LEDER NC1=C(c2ccccc2Cl)c3cc(ccc3NC1=O)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 3-Amino-4-(2-chlorophenyl)-6-nitroquinolin-2(1H)-one
Kvantitet 10 mg
InChI formel InChI=1S/C15H10ClN3O3/c16-11-4-2-1-3-9(11)13-10-7-8(19(21)22)5-6-12(10)18-15(20)14(13)17/h1-7H,17H2,(H,18,20)
Formel vikt 315.0411