missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-fluorobenzoic acid, 97%, Thermo Scientific™

Catalog Number p-7053647
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2365-85-7
C7H6FNO2
155.128
MFCD00130055
WFSPEVFSRUTRCN-UHFFFAOYSA-N
3-amino-4-fluorobenzoicacid, benzoic acid, 3-amino-4-fluoro, 5-carboxy-2-fluoroaniline, 3-amino-4-fluoro-benzoic acid, pubchem4690, acmc-1cilf, ksc207o2p, benzoicacid, 3-amino-4-fluoro, tos-bb-0112, wfspevfsrutrcn-uhfffaoysa
75396
3-amino-4-fluorobenzoic acid
C1=CC(=C(C=C1C(=O)O)N)F
This item is not returnable. View return policy
2365-85-7
C7H6FNO2
155.128
MFCD00130055
WFSPEVFSRUTRCN-UHFFFAOYSA-N
3-amino-4-fluorobenzoicacid, benzoic acid, 3-amino-4-fluoro, 5-carboxy-2-fluoroaniline, 3-amino-4-fluoro-benzoic acid, pubchem4690, acmc-1cilf, ksc207o2p, benzoicacid, 3-amino-4-fluoro, tos-bb-0112, wfspevfsrutrcn-uhfffaoysa
75396
3-amino-4-fluorobenzoic acid
C1=CC(=C(C=C1C(=O)O)N)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 182°C to 184°C
Color White
Quantity 1 g
Formula Weight 155.13
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Amino-4-fluorobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'