Learn More
3-Amino-4-hydroxyphenylboronic acid pinacol ester, 95%, Thermo Scientific™
Catalog Number p-4380040
Click to view available options
Quantity:
1 g
5 g
760990-10-1
C12H18BNO3
235.09
RXHWPAXQYXTPJB-UHFFFAOYSA-N
3-amino-4-hydroxyphenylboronic acid pinacol ester, 2-amino-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-amino-4-hydroxyphenylboronic acid, pinacol ester, 2-amino-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol, 2-amino-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, phenol, 2-amino-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
46738034
2-amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)O)N
760990-10-1
C12H18BNO3
235.09
RXHWPAXQYXTPJB-UHFFFAOYSA-N
3-amino-4-hydroxyphenylboronic acid pinacol ester, 2-amino-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-amino-4-hydroxyphenylboronic acid, pinacol ester, 2-amino-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol, 2-amino-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, phenol, 2-amino-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
46738034
2-amino-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)O)N
Melting Point 151°C to 152°C
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 235.09
Percent Purity 95%
Chemical Name or Material 3-Amino-4-hydroxyphenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 3-Amino-4-hydroxyphenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.