missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-hydroxypyridine hydrate, 95%, Thermo Scientific™

Catalog Number 11322189
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 6320-39-4
Melting Point 145°C to 149°C
Molecular Formula C5H6N2O
MDL Number MFCD04114230
Quantity 1 g
Synonym 3-amino-4-hydroxypyridine, 3-aminopyridin-4-ol, 3-amino-pyridin-4-ol, 4-pyridinol, 3-amino, 3-amino-4-pyridinol, 3-aminopyridin-4 1h-one, 4-hydroxy-3-aminopyridine, 3-amino-4-hydroxypyridine 4-hydroxy-3-amino pyridine, zlchem 466, acmc-209ndv
InChI Key OIIBRAGQGFLUFI-UHFFFAOYSA-N
SMILES NC1=CN=CC=C1O
IUPAC Name 3-aminopyridin-4-ol
Molecular Weight (g/mol) 110.12
PubChem CID 233392
Formula Weight 110.12 (Anhydrous)
Percent Purity 95%
Chemical Name or Material 3-Amino-4-hydroxypyridine hydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'