Learn More
3-Amino-4-methylbenzamide, 98%, Thermo Scientific™
Catalog Number p-56999
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
19406-86-1
C8H10N2O
150.18
MFCD00035936
VYBKAZXQKUFAHG-UHFFFAOYSA-N
benzamide, 3-amino-4-methyl, 3-amino-p-toluamide, 3-amino-4-methyl-benzamide, unii-51vb4677ff, atad, acmc-1c8qe, dsstox_cid_24407, dsstox_rid_80206, 3-amino-4-methyl benzamide, 3-azanyl-4-methyl-benzamide
88043
3-amino-4-methylbenzamide
CC1=C(C=C(C=C1)C(=O)N)N
19406-86-1
C8H10N2O
150.18
MFCD00035936
VYBKAZXQKUFAHG-UHFFFAOYSA-N
benzamide, 3-amino-4-methyl, 3-amino-p-toluamide, 3-amino-4-methyl-benzamide, unii-51vb4677ff, atad, acmc-1c8qe, dsstox_cid_24407, dsstox_rid_80206, 3-amino-4-methyl benzamide, 3-azanyl-4-methyl-benzamide
88043
3-amino-4-methylbenzamide
CC1=C(C=C(C=C1)C(=O)N)N
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 128.0°C to 132.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula NH2C6H3(CH3)CONH2
Quantity 5 g
Formula Weight 150.18
Percent Purity 98%
Physical Form Crystalline Chunks
Chemical Name or Material 3-Amino-4-methylbenzamide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-4-methylbenzamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.