Läs mer
3-Amino-4-propoxybenzoic Acid Hydrochloride, Mikromol™
Produktkod. 17954156
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17954156 100 mg
17944156 25 mg
30461694 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17954156 Leverantör Mikromol Leverantörsnummer MM0404.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Amino-4-propoxybenzoic Acid Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Amino-4-propoxybenzoic Acid Hydrochloride
CAS 59691-14-4
Analyt- eller komponentnamn 3-Amino-4-propoxybenzoic Acid Hydrochloride
Frakt skick Room Temperature
Molekylformel C10 H13 N O3 . Cl H
Certifikat och efterlevnad ISO 17025
Synonym 3-Amino-4-propoxybenzoic Acid Hydrochloride
InChI formel InChI=1S/C10H13NO3.ClH/c1-2-5-14-9-4-3-7(10(12)13)6-8(9)11;/h3-4,6H,2,5,11H2,1H3,(H,12,13);1H
LEDER Cl.CCCOc1ccc(cc1N)C(=O)O
IUPAC-namn 3-amino-4-propoxybenzoic acid;hydrochloride
Molekylvikt (g/mol) 231.68
Kvantitet 100 mg
Formel vikt 231.0662 g/mol