missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-[[[2,3-bis(acetyloxy)propyl]amino]carbonyl]-2,4,6-triiodo-benzoic Acid, TRC

Product Code. 30319481
Change view
Click to view available options
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30319481 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319481 Supplier Toronto Research Chemicals Supplier No. TRCA601970100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 3-Amino-5-(((2,3-bis(acetyloxy)propyl)amino)carbonyl)-2,4,6-triiodo-benzoic Acid
CAS 122731-59-3
Molekylformel C15 H15 I3 N2 O7
Kvantitet 100 mg
Synonym Iopromide Ep Impurity A
InChI formel InChI=1S/C15H15I3N2O7/c1-5(21)26-4-7(27-6(2)22)3-20-14(23)8-10(16)9(15(24)25)12(18)13(19)11(8)17/h7H,3-4,19H2,1-2H3,(H,20,23)(H,24,25)
LEDER CC(=O)OCC(CNC(=O)c1c(I)c(N)c(I)c(C(=O)O)c1I)OC(=O)C
IUPAC-namn 3-amino-5-(2,3-diacetyloxypropylcarbamoyl)-2,4,6-triiodobenzoic acid
Molekylvikt (g/mol) 716.002
Formel vikt 715.801
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.