missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-bromopyridine, 97%, Thermo Scientific™

Catalog Number p-4258744
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
13535-01-8
C5H5BrN2
173.01
MDQXGHBCDCOOSM-UHFFFAOYSA-N
3-amino-5-bromopyridine, 5-bromo-3-pyridinamine, 5-bromo-pyridin-3-ylamine, 5-bromo-3-aminopyridine, 3-pyridinamine, 5-bromo, 5-brompyridin-3-amin, 5-bromo-3-pyridylamine, 5-bromopyridin-3-ylamine, 3-amino-5-bromo pyridine, pubchem1213
817681
5-bromopyridin-3-amine
C1=C(C=NC=C1Br)N
This item is not returnable. View return policy
13535-01-8
C5H5BrN2
173.01
MDQXGHBCDCOOSM-UHFFFAOYSA-N
3-amino-5-bromopyridine, 5-bromo-3-pyridinamine, 5-bromo-pyridin-3-ylamine, 5-bromo-3-aminopyridine, 3-pyridinamine, 5-bromo, 5-brompyridin-3-amin, 5-bromo-3-pyridylamine, 5-bromopyridin-3-ylamine, 3-amino-5-bromo pyridine, pubchem1213
817681
5-bromopyridin-3-amine
C1=C(C=NC=C1Br)N
CAS Min % 96.0
CAS Max % 100.0
Melting Point 62.9°C to 65.2°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 173.01
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Amino-5-bromopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-5-bromopyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'