missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-bromopyridine, 97%, ACROS Organics™

Brand:  Acros Organics 441450010

Additional Details : CAS Number : 13535-01-8 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 11976401

  • 245.00 SEK / 1g

Estimated Shipment: 09-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

13535-01-8
173.01
3-amino-5-bromopyridine, 5-bromo-3-pyridinamine, 5-bromo-pyridin-3-ylamine, 5-bromo-3-aminopyridine, 3-pyridinamine, 5-bromo, 5-brompyridin-3-amin, 5-bromo-3-pyridylamine, 5-bromopyridin-3-ylamine, 3-amino-5-bromo pyridine, pubchem1213
5-bromopyridin-3-amine
C5H5BrN2
MDQXGHBCDCOOSM-UHFFFAOYSA-N
817681
C1=C(C=NC=C1Br)N

Specifications

3-Amino-5-bromopyridine
Glass bottle
62.9°C to 65.2°C
13535-01-8
100.0
C5H5BrN2
MDQXGHBCDCOOSM-UHFFFAOYSA-N
5-bromopyridin-3-amine
817681
Powder
Authentic
Brown to White
1g
96.0
97%
3-amino-5-bromopyridine, 5-bromo-3-pyridinamine, 5-bromo-pyridin-3-ylamine, 5-bromo-3-aminopyridine, 3-pyridinamine, 5-bromo, 5-brompyridin-3-amin, 5-bromo-3-pyridylamine, 5-bromopyridin-3-ylamine, 3-amino-5-bromo pyridine, pubchem1213
C1=C(C=NC=C1Br)N
173.01
173.01
97%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Amino-5-bromopyridine

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents
Certificates