missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-(trifluoromethyl)-1H-indazole, 95%, Thermo Scientific™

Catalog Number p-4228347
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2250-53-5
C8H6F3N3
201.15
MFCD01719740
ZXZTZRVCWTWKCH-UHFFFAOYSA-N
5-trifluoromethyl-1h-indazol-3-amine, 3-amino-5-trifluoromethyl-1h-indazole, 1h-indazol-3-amine, 5-trifluoromethyl, 1h-indazol-3-amine, 5-trifluoromethyl-9ci, 1h-indazole, 3-amino-5-trifluoromethyl, 5-trifluoromethyl-1h-indazole-3-ylamine, 3-amino-5-trifluoromethylindazole, 3-amino-5trifluoromethyl-indazole, 5-trifluoromethyl-1h-indazol-3-ylamine
16736
5-(trifluoromethyl)-1H-indazol-3-amine
C1=CC2=C(C=C1C(F)(F)F)C(=NN2)N
This item is not returnable. View return policy
2250-53-5
C8H6F3N3
201.15
MFCD01719740
ZXZTZRVCWTWKCH-UHFFFAOYSA-N
5-trifluoromethyl-1h-indazol-3-amine, 3-amino-5-trifluoromethyl-1h-indazole, 1h-indazol-3-amine, 5-trifluoromethyl, 1h-indazol-3-amine, 5-trifluoromethyl-9ci, 1h-indazole, 3-amino-5-trifluoromethyl, 5-trifluoromethyl-1h-indazole-3-ylamine, 3-amino-5-trifluoromethylindazole, 3-amino-5trifluoromethyl-indazole, 5-trifluoromethyl-1h-indazol-3-ylamine
16736
5-(trifluoromethyl)-1H-indazol-3-amine
C1=CC2=C(C=C1C(F)(F)F)C(=NN2)N
CAS Min % 94.0
CAS Max % 100.0
Melting Point 134.5°C to 135.5°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 201.15
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-(trifluoromethyl)-1H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-5-(trifluoromethyl)-1H-indazole, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'