Läs mer
3-Amino-6-chloro-4-phenylcarbostyril, TRC
Produktkod. 30317009
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30317009 100 mg
30325653 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30317009 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA603580100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Amino-6-chloro-4-phenylcarbostyril
CAS 5220-83-7
Molekylformel C15 H11 Cl N2 O
Kvantitet 100 mg
Synonym 3-Amino-6-chloro-4-phenyl-carbostyril, 3-Amino-6-chloro-4-phenyl-2-quinolinone
InChI formel InChI=1S/C15H11ClN2O/c16-10-6-7-12-11(8-10)13(14(17)15(19)18-12)9-4-2-1-3-5-9/h1-8H,17H2,(H,18,19)
LEDER NC1=C(c2ccccc2)c3cc(Cl)ccc3NC1=O
IUPAC-namn 3-amino-6-chloro-4-phenyl-1H-quinolin-2-one
Molekylvikt (g/mol) 270.71
Formel vikt 270.056