Läs mer
3-Amino-6-nitro-4-phenylquinolin-2(1H)-one, Mikromol™
Produktkod. 30051703
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051703 25 mg
17970956 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051703 Leverantör Mikromol Leverantörsnummer MM0078.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Amino-6-nitro-4-phenylquinolin-2(1H)-one
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Amino-6-nitro-4-phenylquinolin-2(1H)-one
CAS 36020-93-6
Analyt- eller komponentnamn 3-Amino-6-nitro-4-phenylquinolin-2(1H)-one
Frakt skick Room Temperature
Molekylformel C15 H11 N3 O3
Certifikat och efterlevnad ISO 17025
Synonym 3-Amino-6-nitro-4-phenylquinolin-2(1H)-one, Nitrazepam Imp. A (EP), Nitrazepam Impurity A
InChI formel InChI=1S/C15H11N3O3/c16-14-13(9-4-2-1-3-5-9)11-8-10(18(20)21)6-7-12(11)17-15(14)19/h1-8H,16H2,(H,17,19)
LEDER NC1=C(c2ccccc2)c3cc(ccc3NC1=O)[N+](=O)[O-]
IUPAC-namn 3-amino-6-nitro-4-phenyl-1H-quinolin-2-one
Molekylvikt (g/mol) 281.27
Kvantitet 25 mg
Formel vikt 281.08 g/mol