missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzamidoxime, 97%, Thermo Scientific™

Catalog Number p-7034118
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
100524-07-0
C7H9N3O
151.169
MFCD08061150
OPGWBTKZYLRPRW-UHFFFAOYSA-N
3-aminobenzamidoxime, 3-amino-benzamide oxime, 3-aminobenzamide oxime, 3-amino-n'-hydroxybenzene-1-carboximidamide, z-3-amino-n'-hydroxybenzimidamide, 3-amino-benzamideoxime, 3-aminophenylcarboxamide oxime, 3-amino-n'-hydroxybenzimidamide, 3-amino-n/'-hydroxybenzenecarboximidamide, z-3-amino-n'-hydroxybenzamidine;3-amino-n-hydroxy-benzamidine;3-aminobenzamidoxime
53395946
3-amino-N'-hydroxybenzenecarboximidamide
C1=CC(=CC(=C1)N)C(=NO)N
This item is not returnable. View return policy
100524-07-0
C7H9N3O
151.169
MFCD08061150
OPGWBTKZYLRPRW-UHFFFAOYSA-N
3-aminobenzamidoxime, 3-amino-benzamide oxime, 3-aminobenzamide oxime, 3-amino-n'-hydroxybenzene-1-carboximidamide, z-3-amino-n'-hydroxybenzimidamide, 3-amino-benzamideoxime, 3-aminophenylcarboxamide oxime, 3-amino-n'-hydroxybenzimidamide, 3-amino-n/'-hydroxybenzenecarboximidamide, z-3-amino-n'-hydroxybenzamidine;3-amino-n-hydroxy-benzamidine;3-aminobenzamidoxime
53395946
3-amino-N'-hydroxybenzenecarboximidamide
C1=CC(=CC(=C1)N)C(=NO)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 101°C to 105°C
Quantity 1 g
Formula Weight 151.17
Percent Purity 97%
Chemical Name or Material 3-Aminobenzamidoxime
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'