Läs mer
3-Aminohydantoic Acid, TRC
Produktkod. 30311797
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30311797 1 g
1 options
Produktkod. 30311797 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6111301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Aminohydantoic Acid
CAS 138-07-8
Molekylformel C3H7N3O3
Kvantitet 1 g
Synonym N-Amino-N-(aminocarbonyl)glycine, 1-(Carboxymethyl)hydrazinecarboxamide, N-Amino-N-carbamoylglycine, [1-(Aminocarbonyl)hydrazino]acetic Acid, 2-[1-(Aminocarbonyl)hydrazinyl]acetic Acid
InChI formel InChI=1S/C3H7N3O3/c4-3(9)6(5)1-2(7)8/h1,5H2,(H2,4,9)(H,7,8)
LEDER NN(CC(=O)O)C(=O)N
IUPAC-namn 2-[amino(carbamoyl)amino]acetic acid
Molekylvikt (g/mol) 133.11
Formel vikt 133.0487