missing translation for 'onlineSavingsMsg'
Learn More

3-Aminoisoxazole, 95%, Thermo Scientific Chemicals

Catalog Number p-3777263
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
1750-42-1
C3H4N2O
84.08
MFCD00038814
RHFWLPWDOYJEAL-UHFFFAOYSA-N
3-aminoisoxazole, isoxazol-3-amine, 3-isoxazolamine, 3-amino isoxazole, isoxazolamine, 3-amineisoxazole, aminoisoxazole, 3-aminoisoxazol, 3-amino-1,2-oxazole, amino-isoxazole
1810216
1,2-oxazol-3-amine
C1=CON=C1N
This item is not returnable. View return policy
1750-42-1
C3H4N2O
84.08
MFCD00038814
RHFWLPWDOYJEAL-UHFFFAOYSA-N
3-aminoisoxazole, isoxazol-3-amine, 3-isoxazolamine, 3-amino isoxazole, isoxazolamine, 3-amineisoxazole, aminoisoxazole, 3-aminoisoxazol, 3-amino-1,2-oxazole, amino-isoxazole
1810216
1,2-oxazol-3-amine
C1=CON=C1N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 1.3800g/mL
Boiling Point 226°C to 228°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5100 to 1.5120 (20°C, 589nm)
Quantity 5 mL
Specific Gravity 1.38
Formula Weight 84.08
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Aminoisoxazole
Show More Show Less
compliance-icons
Product Identifier
  • 3-Aminoisoxazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'