missing translation for 'onlineSavingsMsg'
Learn More

3-Aminooxetane, 95%, Thermo Scientific Chemicals

Catalog Number p-7052877
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy
21635-88-1
C3H7NO
73.095
MFCD08544363
OJEOJUQOECNDND-UHFFFAOYSA-N
3-oxetanamine, 3-aminooxetane, oxetan-3-ylamine, 3-oxetanamin, 3-amino-oxetane, oxetane-3-amine, oxetan-3-yl amine, pubchem20414, acmc-1cgam
9833923
oxetan-3-amine
C1C(CO1)N
This item is not returnable. View return policy
21635-88-1
C3H7NO
73.095
MFCD08544363
OJEOJUQOECNDND-UHFFFAOYSA-N
3-oxetanamine, 3-aminooxetane, oxetan-3-ylamine, 3-oxetanamin, 3-amino-oxetane, oxetane-3-amine, oxetan-3-yl amine, pubchem20414, acmc-1cgam
9833923
oxetan-3-amine
C1C(CO1)N

It is used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in medicine.

Solubility
Slightly soluble in water

Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials and heat.
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.05 g/mL
Boiling Point 99°C
Flash Point 24°C (75°F)
Quantity 250 mg
UN Number UN1993
Sensitivity Air sensitive
Solubility Information Slightly soluble in water
Formula Weight 73.09
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Aminooxetane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'