Learn More
3-Aminophenylacetylene, 98%, Thermo Scientific Chemicals
Catalog Number p-7048030
Click to view available options
Quantity:
5 g
25 g
54060-30-9
C8H7N
117.15
MFCD00014779
NNKQLUVBPJEUOR-UHFFFAOYSA-N
3-aminophenylacetylene, benzenamine, 3-ethynyl, m-aminophenylacetylene, 3-ethynylbenzenamine, 3-ethynylphenylamine, 3-ethynylaniline, m-aminophenyl acetylene, 3-aminophenyl acetylene, m-ethynylaniline, 3-ethynyl-aniline
104682
3-ethynylaniline
NC1=CC=CC(=C1)C#C
54060-30-9
C8H7N
117.15
MFCD00014779
NNKQLUVBPJEUOR-UHFFFAOYSA-N
3-aminophenylacetylene, benzenamine, 3-ethynyl, m-aminophenylacetylene, 3-ethynylbenzenamine, 3-ethynylphenylamine, 3-ethynylaniline, m-aminophenyl acetylene, 3-aminophenyl acetylene, m-ethynylaniline, 3-ethynyl-aniline
104682
3-ethynylaniline
NC1=CC=CC(=C1)C#C
It is used as Pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as Pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heavy metal powders.
TRUSTED_SUSTAINABILITY
Melting Point 22°C
Density 1.04
Boiling Point 70°C (1 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.617
Quantity 5 g
Beilstein 2935417
Solubility Information Insoluble in water.
Formula Weight 117.15
Percent Purity 98%
Chemical Name or Material 3-Aminophenylacetylene