missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tocris Bioscience™ 3-AP

Beskrivning
3-AP is a ribonucleotide reductase inhibitor. Also an iron chelator. Inhibits leukemia cell growth and acts in synergy with cisplatin (Cat. No.2251). Active in vivo.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-[(3-Amino-2-pyridinyl)methylene]hydrazinecarbothioamide |
| CAS | 143621-35-6 |
| Synonym | 3-aminopyridine-2-carboxaldehyde thiosemicarbazone |
| Kvantitet | 50 mg |
| Löslighet | Soluble to 100mM in DMSO |
| Molekylformel | C7H9N5S |
| Formel vikt | 195.24 |
| Fysisk form | Solid |
| Färg | White |
| Produkttyp | Bioactive Small Molecule |
| Visa mer |
For Research Use Only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?