Läs mer
3-AP, MedChemExpress
Produktkod. 16495907
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16495907 5 mg
16489726 50 mg
16473178 10 mg
16452656 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16495907 Leverantör Medchem Express Leverantörsnummer HY10082
MedChemExpress 3-AP (PAN-811) is a potent inhibitor of the M2 subunit of ribonucleotide reductase (RR), and is a potent radiosensitizer.
3-AP (PAN-811) is a potent inhibitor of the M2 subunit of ribonucleotide reductase (RR), and is a potent radiosensitizer.
Specifikationer
Kemiskt namn eller material 3-AP
CAS 143621-35-6
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C7H9N5S
Kvantitet 5 mg
Synonym PAN-811 NSC# 663249 OCX191
Löslighetsinformation DMSO : ≥ 47 mg/mL (240.73 mM)
LEDER S=C(N)N/N=C/C1=NC=CC=C1N
Molekylvikt (g/mol) 195.24
Formel vikt 195.24
Procent renhet 99.07%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death