Läs mer
3'-Azido-3'-deoxythymidine 5’-Monophosphate Sodium Salt, TRC
Produktkod. 30340930
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30340930 10 mg
1 options
Produktkod. 30340930 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA82507010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3'-Azido-3'-deoxythymidine 5'-monophosphate sodium salt
CAS 128506-29-6
Anteckningar om renhetsgrad HPLC
Molekylformel C10 H13 N5 O7 P . Na
Kvantitet 10 mg
Synonym 5'-Thymidylic acid, 3'-azido-3'-deoxy-, sodium salt (1:1), 5'-Thymidylic acid, 3'-azido-3'-deoxy-, monosodium salt (9CI), 3'-Azido-3'-deoxythymidine 5'-monophosphate monosodium salt, 3'-Azido-3'-deoxythymidine 5'-phosphate monosodium salt, 3'-Azidothymidine monophosphate monosodium salt, AZT 5'-monophosphate monosodium salt, AZT monophosphate monosodium salt, Azidothymidine monophosphate monosodium salt, Zidovudine monophosphate monosodium salt
InChI formel InChI=1S/C10H14N5O7P.Na/c1-5-3-15(10(17)12-9(5)16)8-2-6(13-14-11)7(22-8)4-21-23(18,19)20;/h3,6-8H,2,4H2,1H3,(H,12,16,17)(H2,18,19,20);/q;+1/p-1/t6-,7+,8+;/m0./s1
LEDER [Na+].CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](COP(=O)(O)[O-])O2)C(=O)NC1=O
IUPAC-namn sodium;[(2S,3S,5R)-3-azido-5-(5-methyl-2,4-dioxopyrimidin-1-yl)oxolan-2-yl]methyl hydrogen phosphate
Molekylvikt (g/mol) 369.2
Formel vikt 369.045