Läs mer
3-Azidopropan-1-amine Hydrochloride Salt, TRC
Produktkod. 30329088
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30329088 100 mg
1 options
Produktkod. 30329088 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA965430100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Azidopropan-1-amine Hydrochloride Salt
CAS 70017-54-8
Molekylformel C3H8N4 . HCl
Kvantitet 100 mg
Synonym 1-Azido-3-aminopropane Monohydrochloride, 3-Azido-1-propanamine Hydrochloride, 3-Azidopropylamine Hydrochloride, N-(3-Azidopropyl)amine Hydrochloride
InChI formel InChI=1S/C3H8N4.ClH/c4-2-1-3-6-7-5;/h1-4H2;1H
LEDER Cl.NCCCN=[N+]=[N-]
IUPAC-namn 3-azidopropan-1-amine;hydrochloride
Molekylvikt (g/mol) 136.58
Formel vikt 136.0516