Läs mer
(3-Azidopropyl)triphenylphosphonium Bromide, TRC
Produktkod. 30009097
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009097 500 mg
30011803 250 mg
30010747 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009097 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA922790500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 127611-39-6
Molekylformel C21 H21 N3 P . Br
Molekylvikt (g/mol) 426.29
Synonym (3-Azidopropyl)triphenyl-phosphonium Bromide
IUPAC-namn 3-azidopropyl(triphenyl)phosphonium;bromide
LEDER [Br-].[N-]=[N+]=NCCC[P+](c1ccccc1)(c2ccccc2)c3ccccc3
Specifikationer
Kemiskt namn eller material (3-Azidopropyl)triphenylphosphonium Bromide
Kvantitet 500 mg
InChI formel InChI=1S/C21H21N3P.BrH/c22-24-23-17-10-18-25(19-11-4-1-5-12-19,20-13-6-2-7-14-20)21-15-8-3-9-16-21;/h1-9,11-16H,10,17-18H2;1H/q+1;/p-1
Formel vikt 425.0656