missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Benzoylbenzoic acid, 98%

943.92 SEK - 2713.78 SEK

Chemical Identifiers

CAS 579-18-0
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD00002518
InChI Key AXJXRLHTQQONQR-UHFFFAOYSA-N
Synonym benzoic acid, 3-benzoyl, m-benzoylbenzoic acid, 3-benzoyl benzoic acid, 3-benzoyl-benzoic acid, 3-phenylcarbonyl benzoic acid, 3-benzoylbenzoicacid, 3-benzoyl-benzoesaure, 3-benzoylbenzoic-acid, acmc-20an9k, benzoic acid,3-benzoyl
PubChem CID 101386
IUPAC Name 3-benzoylbenzoic acid
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11477196 SDS View Documents Alfa Aesar™
B25519.06
5g 943.92 SEK
5g View alternative products
Save Up to 
»
EU Stock 2
Add to basket
 
11487196 SDS View Documents Alfa Aesar™
B25519.14
25g 2713.78 SEK
25g View alternative products
Save Up to 
»
EU Stock 1
Add to basket
 
Description

Description

Applications
3-Benzoylbenzoic acid has been used in light-induced reactions on benzophenone-terminated boron-doped diamond (BDD) surfaces and preparation of benzophenone flavonol derivative.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

579-18-0
226.231
AXJXRLHTQQONQR-UHFFFAOYSA-N
101386
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)C(=O)O
C14H10O3
MFCD00002518
benzoic acid, 3-benzoyl, m-benzoylbenzoic acid, 3-benzoyl benzoic acid, 3-benzoyl-benzoic acid, 3-phenylcarbonyl benzoic acid, 3-benzoylbenzoicacid, 3-benzoyl-benzoesaure, 3-benzoylbenzoic-acid, acmc-20an9k, benzoic acid,3-benzoyl
3-benzoylbenzoic acid

Specifications

3-Benzoylbenzoic acid
162°C to 166°C
579-18-0
MFCD00002518
benzoic acid, 3-benzoyl, m-benzoylbenzoic acid, 3-benzoyl benzoic acid, 3-benzoyl-benzoic acid, 3-phenylcarbonyl benzoic acid, 3-benzoylbenzoicacid, 3-benzoyl-benzoesaure, 3-benzoylbenzoic-acid, acmc-20an9k, benzoic acid,3-benzoyl
AXJXRLHTQQONQR-UHFFFAOYSA-N
3-benzoylbenzoic acid
101386
98%
Odorless
5g
C14H10O3
2212366
Insoluble in water.
C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)C(=O)O
226.231
226.23