Läs mer
3-Benzoyloxyacetophenone, Mikromol™
Produktkod. 30051530
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051530 25 mg
30050972 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051530 Leverantör Mikromol Leverantörsnummer MM0035.500025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Benzoyloxyacetophenone
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-Benzoyloxyacetophenone
CAS 139-28-6
Analyt- eller komponentnamn 3-Benzoyloxyacetophenone
Frakt skick Room Temperature
Molekylformel C15 H12 O3
Certifikat och efterlevnad ISO 17025
Synonym 3-Benzoyloxyacetophenone
InChI formel InChI=1S/C15H12O3/c1-11(16)13-8-5-9-14(10-13)18-15(17)12-6-3-2-4-7-12/h2-10H,1H3
LEDER CC(=O)c1cccc(OC(=O)c2ccccc2)c1
IUPAC-namn (3-acetylphenyl) benzoate
Molekylvikt (g/mol) 240.25
Kvantitet 25 mg
Formel vikt 240.0786 g/mol