missing translation for 'onlineSavingsMsg'
Läs mer

3-Benzoylpyridine, Sigma-Aldrich™

Produktkod. 32238834
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
32238834 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 32238834 Leverantör Sigma Aldrich Leverantörsnummer 8438510025
 Begränsad artikel
Beräknad leverans: 26-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

For synthesis

for synthesis

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 3-Benzoylpyridine
CAS 5424-19-1
Kvantitet 25 g
Synonym 3-Benzoylpyridine, 3-Benzoylpyridine
Molekylvikt (g/mol) 183.21
Kvalitet Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.