Läs mer
3-[Benzyl-(2-methoxycarbonylethyl)amino]propionic Acid Methyl Ester, TRC
Produktkod. 30345598
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30345598 500 mg
1 options
Produktkod. 30345598 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB130175500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-[Benzyl-(2-methoxycarbonylethyl)amino]propionic Acid Methyl Ester
CAS 793-19-1
Molekylformel C15H21NO4
Kvantitet 500 mg
Synonym N-(3-Methoxy-3-oxopropyl)-N-(phenylmethyl)-beta-alanine Methyl Ester, 3,3'-(Benzylimino)dipropionic Acid Dimethyl Ester, Dimethyl 3,3'-(Benzylazanediyl)dipropanoate, N,N-Bis[2-(methoxycarbonyl)ethyl]benzylamine, NSC 54971
InChI formel InChI=1S/C15H21NO4/c1-19-14(17)8-10-16(11-9-15(18)20-2)12-13-6-4-3-5-7-13/h3-7H,8-12H2,1-2H3
LEDER COC(=O)CCN(CCC(=O)OC)Cc1ccccc1
IUPAC-namn methyl 3-[benzyl-(3-methoxy-3-oxopropyl)amino]propanoate
Molekylvikt (g/mol) 279.33
Formel vikt 279.1471