missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Benzyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H64604.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15491879

  • 1783.68 SEK / 1g


Estimated Shipment date
from Supplier 06-08-2018
Add to basket

Specifications

Chemical Identifiers

3-Benzyl-2-thiohydantoin
MFCD00125264
3-Benzyl-2-thioxo-4-imidazolidinone

Specifications

3-Benzyl-2-thiohydantoin
Solid
MFCD00125264
3-Benzyl-2-thioxo-4-imidazolidinone
206.26
95%
1g
Videos
SDS