missing translation for 'onlineSavingsMsg'
Learn More

3-(Benzylamino)propylamine, 95%, Thermo Scientific™

Catalog Number p-7056612
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
13910-48-0
C10H16N2
164.25
MFCD00274361
RFLHDXQRFPJPRR-UHFFFAOYSA-N
n1-benzylpropane-1,3-diamine, 3-benzylamino propylamine, n-benzylpropane-1,3-diamine, 3-aminopropyl benzyl amine, n-benzyl-1,3-propanediamine, n-3-aminopropyl-n-benzylamine, 3-aminopropyl benzylamine, n-benzylpropan-1,3-diamin, n-benzyltrimethylenediamine, n-3-aminopropyl benzylamine
83811
N'-benzylpropane-1,3-diamine
NCCCNCC1=CC=CC=C1
This item is not returnable. View return policy
13910-48-0
C10H16N2
164.25
MFCD00274361
RFLHDXQRFPJPRR-UHFFFAOYSA-N
n1-benzylpropane-1,3-diamine, 3-benzylamino propylamine, n-benzylpropane-1,3-diamine, 3-aminopropyl benzyl amine, n-benzyl-1,3-propanediamine, n-3-aminopropyl-n-benzylamine, 3-aminopropyl benzylamine, n-benzylpropan-1,3-diamin, n-benzyltrimethylenediamine, n-3-aminopropyl benzylamine
83811
N'-benzylpropane-1,3-diamine
NCCCNCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Yellow
Refractive Index 1.532
Quantity 250 mg
Beilstein 509316
Formula Weight 164.25
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-(Benzylamino)propylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'